ENA155457785 Enamine

ethyl (2E)-2-cyano-3-thien-2-ylbut-2-enoate

  • MDL number MFCD09863437
  • Cas Number : 201733-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30875

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30875
Availability details for
Quantity
  • Availability
  • Add to Cart