ENA155458064 Enamine

3-(morpholin-4-yl)-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD03447775
  • Cas Number : 18630-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30816
Availability details for
Quantity
  • Availability
  • Add to Cart