ENA155458087 Enamine

1H,2H,3H,4H-cyclopenta[b]indole-5-carboxylic acid

  • MDL number MFCD09863427
  • Cas Number : 422312-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30836
Availability details for
Quantity
  • Availability
  • Add to Cart