ENA155458102 Enamine

1-(4-fluorophenyl)-2-(phenylthio)ethanone

  • MDL number MFCD03485960
  • Cas Number : 107820-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30857

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30857
Availability details for
Quantity
  • Availability
  • Add to Cart