ENA155458106 Enamine

3-(3-chloropropanoyl)-1-(2,3-dihydro-1,4-benzodioxin-6-yl)urea

  • MDL number MFCD09863266
  • Cas Number : 1000930-40-4
  • Molecular Weight : 284.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-25857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25857
Availability details for
Quantity
  • Availability
  • Add to Cart