ENA155458108 Enamine

N-(1-benzylpiperidin-4-yl)-1H-imidazole-1-carboxamide

  • MDL number MFCD09863435
  • Cas Number : 380222-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30867
Availability details for
Quantity
  • Availability
  • Add to Cart