ENA155458119 Enamine

1-(3-methylbenzoyl)-1,4-diazepane

  • MDL number MFCD05879206
  • Cas Number : 815650-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30888

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30888
Availability details for
Quantity
  • Availability
  • Add to Cart