ENA155458123 Enamine

N-(3-morpholin-4-ylpropyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD08443468
  • Cas Number : 926246-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182...184
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30893

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30893
Availability details for
Quantity
  • Availability
  • Add to Cart