ENA155458133 Enamine

3-chloro-1-[4-(pyridin-2-yl)piperazin-1-yl]propan-1-one

  • MDL number MFCD09808364
  • Cas Number : 952482-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-30915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30915
Availability details for
Quantity
  • Availability
  • Add to Cart