ENA155458134 Enamine

3-chloro-1-[4-(4-fluorophenyl)piperazin-1-yl]propan-1-one

  • MDL number MFCD00784254
  • Cas Number : 251638-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-30916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30916
Availability details for
Quantity
  • Availability
  • Add to Cart