ENA155458136 Enamine

[(1-methyl-1H-indol-3-yl)methylene]malononitrile

  • MDL number MFCD01232331
  • Cas Number : 65037-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30922

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30922
Availability details for
Quantity
  • Availability
  • Add to Cart