ENA155458143 Enamine

5-(4-chloro-3-nitrophenyl)-2-furaldehyde

  • MDL number MFCD01152623
  • Cas Number : 714923-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30934

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30934
Availability details for
Quantity
  • Availability
  • Add to Cart