ENA155458149 Enamine

2-(1,4-diazepan-1-yl)-N,N-diethylacetamide

  • MDL number MFCD09050136
  • Cas Number : 87055-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30959
Availability details for
Quantity
  • Availability
  • Add to Cart