ENA155458153 Enamine

2-(2-bromoethyl)-2H,3H-[1,2,4]triazolo[4,3-a]pyridin-3-one

  • MDL number MFCD09803820
  • Cas Number : 1000932-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30970
Availability details for
Quantity
  • Availability
  • Add to Cart