ENA155458160 Enamine

methyl 5-(cyanomethyl)furan-2-carboxylate

  • MDL number MFCD03361694
  • Cas Number : 39258-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30977
Availability details for
Quantity
  • Availability
  • Add to Cart