ENA155458364 Enamine

3-methoxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzoic acid

  • MDL number MFCD08445235
  • Cas Number : 923697-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30804
Availability details for
Quantity
  • Availability
  • Add to Cart