ENA155458497 Enamine

2-ethoxypyridine-3-carbonitrile

  • MDL number MFCD08436064
  • Cas Number : 14248-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30814
Availability details for
Quantity
  • Availability
  • Add to Cart