ENA156508039 Enamine

4-(chloromethyl)-2-(3-chlorophenyl)-1,3-oxazole

  • MDL number MFCD09971345
  • Cas Number : 22091-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29756
Availability details for
Quantity
  • Availability
  • Add to Cart