ENA156508046 Enamine

4-(trifluoromethyl)-12-oxa-2,3,5,7-tetraazatricyclo[7.4.0.0,2,6]trideca-1(9),3,5,7-tetraen-13-one

  • MDL number MFCD09971381
  • Cas Number : 1000932-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31038
Availability details for
Quantity
  • Availability
  • Add to Cart