ENA156508054 Enamine

N-1,3-benzothiazol-2-ylpyrrolidine-2-carboxamide hydrochloride

  • MDL number MFCD09971410
  • Cas Number : 1078162-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31170

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31170
Availability details for
Quantity
  • Availability
  • Add to Cart