ENA156508065 Enamine

2-(dimethylamino)-1-(piperazin-1-yl)ethan-1-one dihydrochloride

  • MDL number MFCD09971434
  • Cas Number : 1177336-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31247
Availability details for
Quantity
  • Availability
  • Add to Cart