ENA156508076 Enamine

1-[1-(4-fluorophenyl)-5-methyl-1H-pyrazol-4-yl]ethanamine hydrochloride

  • MDL number MFCD09971462
  • Cas Number : 1172969-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31335

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31335
Availability details for
Quantity
  • Availability
  • Add to Cart