ENA156508077 Enamine

1-[5-methyl-1-(4-methylphenyl)-1H-pyrazol-4-yl]ethan-1-amine hydrochloride

  • MDL number MFCD09971464
  • Cas Number : 1172228-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-31337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31337
Availability details for
Quantity
  • Availability
  • Add to Cart