ENA156508085 Enamine

4-allyl-5-amino-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD09971476
  • Cas Number : 76267-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31392

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31392
Availability details for
Quantity
  • Availability
  • Add to Cart