ENA156508091 Enamine

5-nitro-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD09971490
  • Cas Number : 13138-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31446

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31446
Availability details for
Quantity
  • Availability
  • Add to Cart