ENA156508097 Enamine

4-amino-1H-pyrrole-2-carboxamide hydrochloride

  • MDL number MFCD09971498
  • Cas Number : 1172552-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31490

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31490
Availability details for
Quantity
  • Availability
  • Add to Cart