ENA156508121 Enamine

2-nitro-5H,6H,7H,8H,9H,10H-cyclohepta[b]indole

  • MDL number MFCD09971542
  • Cas Number : 17993-55-4
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-31702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31702
Availability details for
Quantity
  • Availability
  • Add to Cart