ENA156508169 Enamine

1-(pyridin-3-ylcarbonyl)piperidine-4-carboxylic acid

  • MDL number MFCD00434437
  • Cas Number : 67691-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188...190
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31113

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31113
Availability details for
Quantity
  • Availability
  • Add to Cart