ENA156508170 Enamine

6-sulfanylpyridine-3-carbonitrile

  • MDL number MFCD09152765
  • Cas Number : 27885-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253-255
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31114
Availability details for
Quantity
  • Availability
  • Add to Cart