ENA156508172 Enamine

1-(cyclopropylcarbonyl)piperidin-4-one oxime

  • MDL number MFCD09931489
  • Cas Number : 1000932-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31118

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31118
Availability details for
Quantity
  • Availability
  • Add to Cart