ENA156508173 Enamine

1-(ethanesulfonyl)piperidine-3-carboxylic acid

  • MDL number MFCD03830441
  • Cas Number : 702695-86-1
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-31119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31119
Availability details for
Quantity
  • Availability
  • Add to Cart