ENA156508176 Enamine

3,5-bis(methylthio)-1,2,4-thiadiazole

  • MDL number MFCD01935194
  • Cas Number : 41218-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31127

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31127
Availability details for
Quantity
  • Availability
  • Add to Cart