ENA156508177 Enamine

amino(pyridin-3-yl)acetonitrile

  • MDL number MFCD09042089
  • Cas Number : 131988-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31132

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31132
Availability details for
Quantity
  • Availability
  • Add to Cart