ENA156508188 Enamine

2-(1-cyclopropyl-N-methylformamido)acetic acid

  • MDL number MFCD09050622
  • Cas Number : 926213-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31146
Availability details for
Quantity
  • Availability
  • Add to Cart