ENA156508189 Enamine

3-(cyclopropylformamido)propanoic acid

  • MDL number MFCD07428852
  • Cas Number : 874455-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31147
Availability details for
Quantity
  • Availability
  • Add to Cart