ENA156508192 Enamine

ethyl 6-piperazin-1-ylnicotinate

  • MDL number MFCD00894968
  • Cas Number : 132521-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31151

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31151
Availability details for
Quantity
  • Availability
  • Add to Cart