ENA156508194 Enamine

5-oxo-1-(3,4,5-trimethoxyphenyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD08443125
  • Cas Number : 926190-67-2
  • Molecular Weight : 295.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-31153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31153
Availability details for
Quantity
  • Availability
  • Add to Cart