ENA156508204 Enamine

1-(4-chlorophenyl)-5-isopropyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09971409
  • Cas Number : 187998-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31168

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31168
Availability details for
Quantity
  • Availability
  • Add to Cart