ENA156508208 Enamine

2-chloro-N-(quinolin-5-yl)propanamide

  • MDL number MFCD09945534
  • Cas Number : 1000930-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28015
Availability details for
Quantity
  • Availability
  • Add to Cart