ENA156508210 Enamine

3-(aminomethyl)-N-(propan-2-yl)benzene-1-sulfonamide

  • MDL number MFCD09735338
  • Cas Number : 953715-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31176
Availability details for
Quantity
  • Availability
  • Add to Cart