ENA156508212 Enamine

N-[3-(1-aminoethyl)phenyl]acetamide

  • MDL number MFCD09738445
  • Cas Number : 918810-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31179
Availability details for
Quantity
  • Availability
  • Add to Cart