ENA156508213 Enamine

3-(cyclopentyloxy)-4-methoxyaniline hydrochloride

  • MDL number MFCD09971414
  • Cas Number : 844873-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31180
Availability details for
Quantity
  • Availability
  • Add to Cart