ENA156508214 Enamine

3-cyanobenzamide

  • MDL number MFCD00017574

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31182
Availability details for
Quantity
  • Availability
  • Add to Cart