ENA156508215 Enamine

2-(2-methoxyacetamido)propanoic acid

  • MDL number MFCD09046076
  • Cas Number : 257638-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31183
Availability details for
Quantity
  • Availability
  • Add to Cart