ENA156508225 Enamine

1-(4-fluorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD02677737
  • Cas Number : 217073-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29372

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29372
Availability details for
Quantity
  • Availability
  • Add to Cart