ENA156508226 Enamine

5-(4-ethoxyphenyl)-2H-1,2,3,4-tetrazole

  • MDL number MFCD02739910
  • Cas Number : 338426-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31200
Availability details for
Quantity
  • Availability
  • Add to Cart