ENA156508230 Enamine

N-cyclopropylcyclopentanamine

  • MDL number MFCD09048840
  • Cas Number : 873-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31207
Availability details for
Quantity
  • Availability
  • Add to Cart