ENA156508232 Enamine

1H,2H,3H,4H,5H-pyrido[4,3-b]indole-6-carboxylic acid hydrochloride

  • MDL number MFCD09971422
  • Cas Number : 444721-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31211
Availability details for
Quantity
  • Availability
  • Add to Cart