ENA156508234 Enamine

2-[(ethylsulfonyl)amino]propanoic acid

  • MDL number MFCD09048314

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31213

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31213
Availability details for
Quantity
  • Availability
  • Add to Cart