ENA156508241 Enamine

N,2-dimethylcyclohexanamine hydrochloride

  • MDL number MFCD09971428
  • Cas Number : 101257-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31224

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31224
Availability details for
Quantity
  • Availability
  • Add to Cart